missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetic acid, 98.5%

Product Code. 10316970 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Unit Size:
2.5kg
500g
5g
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Product Code. 10316970

Brand: Thermo Scientific Acros 130285000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

CAS: 103-82-2 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-82-2
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
InChI Key WLJVXDMOQOGPHL-UHFFFAOYSA-N
IUPAC Name 2-phenylacetic acid
SMILES OC(=O)CC1=CC=CC=C1

Specifications

Melting Point 75.0°C to 78.0°C
Density 1.0800 g/mL
Boiling Point 265.0°C
Flash Point 132°C
Assay Percent Range 98.5%
Packaging Plastic bottle
Linear Formula C6H5CH2CO2H
Quantity 500 g
Beilstein 09, 431
Merck Index 15, 7380
Specific Gravity 1.08
Solubility Information Solubility aq. soln.: 15 g/L (20°C). Other solubilities: freely soluble in hot water, soluble in ether, acetone, 186 g/100 mL ethanol (20°C), 4.422 mol.s/l in chloroform (25°C), 1.842 mol.s/l in carbon tetrachloride
Formula Weight 136.15
Chemical Name or Material Phenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • Phenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.