missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylacetic acid, 98.5%, Thermo Scientific Chemicals
14.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 75.0°C to 78.0°C |
Density | 1.0800 g/mL |
Boiling Point | 265.0°C |
Flash Point | 132°C |
Assay Percent Range | 98.5% |
Packaging | Glass bottle |
Linear Formula | C6H5CH2CO2H |
Quantity | 5 g |
Beilstein | 09, 431 |
Merck Index | 15, 7380 |
Show More |
Safety and Handling
Product Identifier
- Phenylacetic acid
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'