missing translation for 'onlineSavingsMsg'
Learn More

Phenyl sulfide, 99%, Thermo Scientific Chemicals

Catalog Number p-184108
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
139-66-2
C12H10S
186.27
MFCD00003064
LTYMSROWYAPPGB-UHFFFAOYSA-N
diphenyl sulfide, phenyl sulfide, phenylthiobenzene, diphenyl sulphide, diphenyl thioether, benzene, 1,1'-thiobis, diphenylmercaptan, diphenylthiamethane, diphenyl monosulfide, sulfide, diphenyl
8766
CHEBI:38959
phenylsulfanylbenzene
S(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
139-66-2
C12H10S
186.27
MFCD00003064
LTYMSROWYAPPGB-UHFFFAOYSA-N
diphenyl sulfide, phenyl sulfide, phenylthiobenzene, diphenyl sulphide, diphenyl thioether, benzene, 1,1'-thiobis, diphenylmercaptan, diphenylthiamethane, diphenyl monosulfide, sulfide, diphenyl
8766
CHEBI:38959
phenylsulfanylbenzene
S(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -40°C
Color Colorless to Yellow
Density 1.1100g/mL
Boiling Point 296°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C6H5)2S
Refractive Index 1.6317 to 1.6337
Quantity 25 mL
Beilstein 06, 299
Fieser 06,241
Merck Index 15, 3371
Specific Gravity 1.11
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in hot alcohol,miscible with benzene,ether and carbon disulfide
Formula Weight 186.27
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenyl sulfide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Phenyl sulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'