missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals
68.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Phenyl phosphate disodium salt is used in phosphatase determinations. It is also used as reagent in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Linear Formula | C6H5OPO(ONa)2·xH2O |
Quantity | 25 g |
Beilstein | 4167154 |
Sensitivity | Hygroscopic |
Merck Index | 14,3357 |
Solubility Information | Soluble in water (0.1g/mL). |
Formula Weight | 218.06 (Anhydrous) |
Percent Purity | 98% |
Physical Form | Crystalline |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'