missing translation for 'onlineSavingsMsg'
Learn More

Phenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7081570
Click to view available options
Quantity:
5 g
100 g
500 g
This item is not returnable. View return policy
103-72-0
C7H5NS
135.18
MFCD00004798
QKFJKGMPGYROCL-UHFFFAOYSA-N
phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
7673
CHEBI:85103
isothiocyanatobenzene
S=C=NC1=CC=CC=C1
This item is not returnable. View return policy
103-72-0
C7H5NS
135.18
MFCD00004798
QKFJKGMPGYROCL-UHFFFAOYSA-N
phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
7673
CHEBI:85103
isothiocyanatobenzene
S=C=NC1=CC=CC=C1

Phenyl isothiocyanate acts as a derivatizing reagent for primary and secondary amines. It is used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC. It is used for derivatizing N-terminal amino acids of proteins for automated sequential analysis. It is a synthon for dithiadiazafulvalenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl isothiocyanate acts as a derivatizing reagent for primary and secondary amines. It is used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC. It is used for derivatizing N-terminal amino acids of proteins for automated sequential analysis. It is a synthon for dithiadiazafulvalenes.

Solubility
Soluble in alcohol, and ether. Insoluble in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Store at 4°C.
TRUSTED_SUSTAINABILITY
Melting Point -21°C
Density 1.131
Boiling Point 220°C to 221°C
Flash Point 87°C (188°F)
Odor Pungent
Refractive Index 1.65
Quantity 5 g
UN Number UN2927
Beilstein 471392
Sensitivity Moisture sensitive
Merck Index 14,7297
Solubility Information Soluble in alcohol,and ether. Insoluble in water.
Formula Weight 135.19
Percent Purity 97%
Chemical Name or Material Phenyl isothiocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'