missing translation for 'onlineSavingsMsg'
Learn More

γ-Phenyl-γ-butyrolactone, 97%, Thermo Scientific™

Catalog Number 10488320
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1008-76-0
C10H10O2
162.19
MFCD00005398
AEUULUMEYIPECD-UHFFFAOYNA-N
gamma-phenyl-gamma-butyrolactone, 4-phenylbutanolide, 4-phenyl-4-butanolide, 4-phenylbutyrolactone, 2 3h-furanone, dihydro-5-phenyl, 5-phenyldihydrofuran-2 3h-one, 4-phenyl-4-hydroxybutanoic acid lactone, 4,5-dihydro-5-phenyl-2 3h-furanone, .gamma.-phenyl-.gamma.-butyrolactone, .gamma.-phenylbutyrolactone
13884
5-phenyloxolan-2-one
O=C1CCC(O1)C1=CC=CC=C1
This item is not returnable. View return policy
1008-76-0
C10H10O2
162.19
MFCD00005398
AEUULUMEYIPECD-UHFFFAOYNA-N
gamma-phenyl-gamma-butyrolactone, 4-phenylbutanolide, 4-phenyl-4-butanolide, 4-phenylbutyrolactone, 2 3h-furanone, dihydro-5-phenyl, 5-phenyldihydrofuran-2 3h-one, 4-phenyl-4-hydroxybutanoic acid lactone, 4,5-dihydro-5-phenyl-2 3h-furanone, .gamma.-phenyl-.gamma.-butyrolactone, .gamma.-phenylbutyrolactone
13884
5-phenyloxolan-2-one
O=C1CCC(O1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 36.0°C to 37.0°C
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 306.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5415 to 1.5435
Quantity 5 g
Beilstein 17,319
Specific Gravity 1.15
Formula Weight 162.19
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material γ-Phenyl-γ-butyrolactone, 97%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'