missing translation for 'onlineSavingsMsg'
Learn More

Phenoxyacetic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-4510518
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
122-59-8
C8H8O3
152.15
MFCD00004296
LCPDWSOZIOUXRV-UHFFFAOYSA-N
phenoxyacetic acid, phenoxyethanoic acid, acetic acid, phenoxy, glycolic acid phenyl ether, phenoxy acetic acid, glycol acid phenyl ether, o-phenylglycolic acid, phenoxyacetate, glycollic acid phenyl ether, acide phenoxyacetique
19188
CHEBI:8075
2-phenoxyacetic acid
OC(=O)COC1=CC=CC=C1
This item is not returnable. View return policy
122-59-8
C8H8O3
152.15
MFCD00004296
LCPDWSOZIOUXRV-UHFFFAOYSA-N
phenoxyacetic acid, phenoxyethanoic acid, acetic acid, phenoxy, glycolic acid phenyl ether, phenoxy acetic acid, glycol acid phenyl ether, o-phenylglycolic acid, phenoxyacetate, glycollic acid phenyl ether, acide phenoxyacetique
19188
CHEBI:8075
2-phenoxyacetic acid
OC(=O)COC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 98.0°C to 101.0°C
Color White
Boiling Point 285.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (Acid)
Packaging Glass Bottle
Linear Formula C6H5OCH2CO2H
Quantity 5 g
Beilstein 06,161
Fieser 09,361
Merck Index 15,7366
Solubility Information Solubility in water: soluble. Other solubilities: soluble in glacial acetic acid, soluble in ether, alcohol and carbon disulfide
Formula Weight 152.15
Percent Purity 98+%
Physical Form Granular Powder, Granules, or Prills
Chemical Name or Material Phenoxyacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • Phenoxyacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'