missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Solid, Pure, Indicator Grade, Fisher Chemical™

Catalog Number p-7073389
35.76 GBP valid until 2024-12-31
Use promo code "21984" to get your promotional price.


35.76 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


49.12 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
77-09-8
C20H14O4
318.33
MFCD00005913
KJFMBFZCATUALV-UHFFFAOYSA-N
4764
CHEBI:34914
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
This item is not returnable. View return policy
77-09-8
C20H14O4
318.33
MFCD00005913
KJFMBFZCATUALV-UHFFFAOYSA-N
4764
CHEBI:34914
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Phenolphthalein Solid, Pure, Indicator Grade

TRUSTED_SUSTAINABILITY
Melting Point 258°C
Formula Weight 318.32g/mol
Packaging Plastic powder jar
Color White
Quantity 100 g
Physical Form Solid
compliance-icons
Product Identifier
  • Phenolphthalein
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1
  • Reproductive toxicity Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : phenolphthalein
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'