missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Phenolphthalein, 0.5% w/v in alcohol
48.55 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Use promo code "21687" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Formula Weight | 318.33 |
Odor | Alcohol-like |
Density | 0.918 g/cm3 at 20°C |
Color | Colorless |
Quantity | 500 mL |
Flash Point | 11°C (52°F) |
Physical Form | Liquid |
Chemical Name or Material | Phenolphthalein |
For Manufacturing and Laboratory Use. Not for beverage use
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'