missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red, Sodium Salt, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5391558
280.25 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
34487-61-1
C19H13NaO5S
376.358
HKHYOKBQJILTEI-UHFFFAOYSA-M
23686673
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
This item is not returnable. View return policy
34487-61-1
C19H13NaO5S
376.358
HKHYOKBQJILTEI-UHFFFAOYSA-M
23686673
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 376.37
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Quantity 5 g
Physical Form Powder
Chemical Name or Material Phenol Red, Sodium Salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'