missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red, Sodium Salt, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
280.25 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Formula Weight | 376.37 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | Phenol Red, Sodium Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'