missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Phenol, Ready-to-Use, buffer saturated solution pH 4.5

Brand:  Alfa Aesar™ J67158.AE

Code : NEW

Additional Details : Weight : 0.00010kg Transport : UN number : 2821 Chem class : 6.1 Pack group : II

Quantity 100mL

 View more versions of this product

Product Code. 15410717

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Phenol
Ready-to-Use, buffer saturated solution
MFCD00002143
6428463
108-95-2
94.11
UN2821

Specifications

Phenol
108-95-2
94.11
MFCD00002143
6428463
4.5
Light Sensitive
Ready-to-Use, buffer saturated solution
Liquid
1.06g/mL
UN2821
Clear
100mL
Videos
SDS