missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Phenol, Ready-to-Use, buffer saturated solution pH 4.5

Brand:  Alfa Aesar™ J67158.AN

Code : NEW

Additional Details : Weight : 0.00040kg Transport : UN number : 2821 Chem class : 1 Pack group : II

Quantity 400mL

 View more versions of this product

Product Code. 15420717

  • £94.50 / 400mL


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

Phenol
Ready-to-Use, buffer saturated solution
MFCD00002143
6428463
108-95-2
94.11
UN2821

Specifications

Phenol
108-95-2
94.11
MFCD00002143
6428463
4.5
Light Sensitive
Ready-to-Use, buffer saturated solution
Liquid
1.06g/mL
UN2821
Clear
400mL
Videos
SDS