missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Honeywell Fluka™

Catalog Number p-7145726
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
108-95-2
C6H6O
94.11
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
996
CHEBI:15882
phenol
OC1=CC=CC=C1
This item is not returnable. View return policy
108-95-2
C6H6O
94.11
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
996
CHEBI:15882
phenol
OC1=CC=CC=C1

Puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Acidity Complying
Flash Point 79 °C
Packaging Plastic Bottle
Quantity 500 g
Percent Purity 99.5 - 100.5 %
Grade Reagent Pharmacopeia Organics
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'