Phenethylamine, 99%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 156490010

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 64-04-0 Weight: 1.70000kg Transport: UN number : 2735 Chem class : 1b Pack group : I


Packaging Glass bottle
Quantity 1kg

Product Code. 10605212

Quantity Price
1 £ 59.1 / 1kg
EU Stock 1
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Description and Specification

Chemical Identifiers

Chemical Name or Material Phenethylamine
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H11N
Linear Formula C6H5CH2CH2NH2
Formula Weight 121.18
MDL Number MFCD00008184
Synonym 2-Phenylethylamine

Specification

Assay ≥98.5%
Physical Form Liquid
Formula Weight 121.18
Quantity 1kg
Color Clear colorless to pale yellow
Fieser 09,363
Beilstein 12, 1096
Merck Index 15, 7339
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula C6H5CH2CH2NH2
Infrared Spectrum Authentic
CAS 64-04-0
Assay Percent Range 99%
Refractive Index 1.5320 to 1.5340 (20°C, 589nm)
MDL Number MFCD00008184
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Packaging Glass bottle
Water 0.5% max.
Health Hazard 1 Danger
Molecular Formula C8H11N
Density 0.9620g/mL
Chemical Name or Material Phenethylamine
Synonym 2-Phenylethylamine
Documentation

Technical documents

Safety and Handling

Danger

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.