Phenethylamine, 99%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 156491000

100GR Phenethylamine, 99%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 64-04-0 Weight: 0.15000kg Transport: UN number : 2735 Chem class : 1b Pack group : I


Packaging Glass bottle
Quantity 100g

Product Code. 10286200

Quantity Price
1 £ 10.3 / 100g
EU Stock 11
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Phenethylamine
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H11N
Linear Formula C6H5CH2CH2NH2
Formula Weight 121.18
MDL Number MFCD00008184
Synonym 2-Phenylethylamine

Specification

Assay ≥98.5%
Physical Form Liquid
Formula Weight 121.18
Quantity 100g
Color Clear colorless to pale yellow
Beilstein 12, 1096
Fieser 09,363
Merck Index 15, 7339
Linear Formula C6H5CH2CH2NH2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 64-04-0
Assay Percent Range 99%
Refractive Index 1.5320 to 1.5340 (20°C, 589nm)
MDL Number MFCD00008184
Packaging Glass bottle
Water 0.5% max.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Molecular Formula C8H11N
Health Hazard 1 Danger
Density 0.9620g/mL
Chemical Name or Material Phenethylamine
Synonym 2-Phenylethylamine
Documentation

Technical documents

Safety and Handling

Danger

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.