missing translation for 'onlineSavingsMsg'
Learn More

Phenazine methosulfate, 98%, Thermo Scientific Chemicals

Catalog Number 10785581
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
299-11-6
C13H11N2·CH3O4S
306.33
MFCD00011923
RXGJTUSBYWCRBK-UHFFFAOYSA-M
phenazine methosulfate, 5-methylphenazin-5-ium methyl sulfate, 5-methylphenazinium methyl sulfate, n-methylphenazonium methosulfate, phenazine methosulphate, methylphenazonium methosulfate, 5-methylphenazine methylsulfate, n-methylphenazinium methosulfate, n-methylphenazonium methosulphate, phenazinium, 5-methyl-, methyl sulfate
9285
CHEBI:8055
5-methylphenazin-5-ium;methyl sulfate
C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-]
This item is not returnable. View return policy
299-11-6
C13H11N2·CH3O4S
306.33
MFCD00011923
RXGJTUSBYWCRBK-UHFFFAOYSA-M
phenazine methosulfate, 5-methylphenazin-5-ium methyl sulfate, 5-methylphenazinium methyl sulfate, n-methylphenazonium methosulfate, phenazine methosulphate, methylphenazonium methosulfate, 5-methylphenazine methylsulfate, n-methylphenazinium methosulfate, n-methylphenazonium methosulphate, phenazinium, 5-methyl-, methyl sulfate
9285
CHEBI:8055
5-methylphenazin-5-ium;methyl sulfate
C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 158°C to 160°C
Color Brown to Yellow or Khaki
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Merck Index 15, 6182
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol and ethanol,insoluble in benzene and diethylether
Formula Weight 306.33
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Phenazine methosulfate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenazine methosulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'