missing translation for 'onlineSavingsMsg'
Learn More

Phenazine, 98%, Thermo Scientific Chemicals

Catalog Number 15199173
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
92-82-0
C12H8N2
180.21
MFCD00005023
PCNDJXKNXGMECE-UHFFFAOYSA-N
dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs
4757
CHEBI:36674
phenazine
C1=CC=C2C(=C1)N=C3C=CC=CC3=N2
This item is not returnable. View return policy
92-82-0
C12H8N2
180.21
MFCD00005023
PCNDJXKNXGMECE-UHFFFAOYSA-N
dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs
4757
CHEBI:36674
phenazine
C1=CC=C2C(=C1)N=C3C=CC=CC3=N2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 174.0°C to 177.0°C
Color Yellow to Brown
Boiling Point >360.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 23,223
Merck Index 15,7331
Solubility Information Solubility in water: insoluble. Other solubilities: partially soluble in alcohol and ether, moderately soluble in benzene
Formula Weight 180.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Phenazine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'