missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene, 98%, Thermo Scientific Chemicals

Catalog Number p-7043656
Click to view available options
Quantity:
50 g
250 g
85-01-8
C14H10
178.23
MFCD00001168
YNPNZTXNASCQKK-UHFFFAOYSA-N
phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730
995
CHEBI:28851
phenanthrene
C1=CC=C2C(C=CC3=CC=CC=C23)=C1
85-01-8
C14H10
178.23
MFCD00001168
YNPNZTXNASCQKK-UHFFFAOYSA-N
phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730
995
CHEBI:28851
phenanthrene
C1=CC=C2C(C=CC3=CC=CC=C23)=C1

Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.

Solubility
Soluble in Ether, Benzene, Chloroform, Toluene. Insoluble in water.

Notes
Light sensitive. Handle and store under inert gas. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 97°C to 101°C
Density 1.179
Boiling Point 340°C
Flash Point 171°C (339°F)
Quantity 50 g
UN Number UN3077
Beilstein 1905428
Merck Index 14,7212
Solubility Information Soluble in Ether,Benzene,Chloroform,Toluene. Insoluble in water.
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material Phenanthrene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'