missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ [Phe&superscript_7;] Dynorphin A (1-7), Porcine

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J65474.MB

UNSPSC : 12352202

Code : NEW

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15436919

  • £235.64 / 25mg


Estimated Shipment date
from Supplier 07-08-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

25mg
[Phe7] Dynorphin A (1-7), porcine
858.98
RUO
Porcine
Tyr-Gly-Gly-Phe-Leu-Arg-Phe
Solid
HPLC
-30° to -10°C
Documentation

Technical Documents