Learn More
Phaclofen, Tocris Bioscience™
Selective GABAB antagonist
Brand:  Tocris (Bio-Techne) 017810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13467453
  • £111.72 / 10mg
Estimated Shipment date
from Supplier 26-03-2020
Description
Description
Selective GABAB antagonist. Phosphono analog of GABAB agonist baclofen (Cat. No. 0417).
Specifications
Chemical Identifiers
114012-12-3
249.631
phaclofen, 3-amino-2-4-chlorophenyl propyl phosphonic acid, phosphonic acid, 3-amino-2-4-chlorophenyl propyl, phosphonic acid,p-3-amino-2-4-chlorophenyl propyl, 3-amino-2-4-chlorophenyl propylphosphonic acid, phaclofen, solid, beta-4-chlorophenyl-3-aminopropylphosphonic acid, acmc-20djlx, biomol-nt_000256
[3-amino-2-(4-chlorophenyl)propyl]phosphonic acid
C9H13ClNO3P
VSGNGLJPOGUDON-UHFFFAOYSA-N
1641
C1=CC(=CC=C1C(CN)CP(=O)(O)O)Cl
Specifications
Phaclofen
114012-12-3
phaclofen, 3-amino-2-4-chlorophenyl propyl phosphonic acid, phosphonic acid, 3-amino-2-4-chlorophenyl propyl, phosphonic acid,p-3-amino-2-4-chlorophenyl propyl, 3-amino-2-4-chlorophenyl propylphosphonic acid, phaclofen, solid, beta-4-chlorophenyl-3-aminopropylphosphonic acid, acmc-20djlx, biomol-nt_000256
C1=CC(=CC=C1C(CN)CP(=O)(O)O)Cl
249.631
249.63
10mg
C9H13ClNO3P
VSGNGLJPOGUDON-UHFFFAOYSA-N
[3-amino-2-(4-chlorophenyl)propyl]phosphonic acid
1641
>97%