missing translation for 'onlineSavingsMsg'
Learn More

PHA 767491 hydrochloride, Tocris Bioscience™

MK2 inhibitor; also cdk inhibitor; potently inhibits cdk9

Brand:  Tocris (Bio-Techne) 314050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13442514

  • £573.30 / 50mg

Estimated Shipment date
from Supplier 28-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ATP-competitive cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 0.034 - 0.75, 0.24 - 1.2, 0.25 - 1.1 and 0.46 - 1 μM for cdk9, cdk2, cdk1 and cdk5, respectively). Also inhibits mitogen-activated protein kinase-activated protein kinase-2 (MK2; IC50 = 171 nM). Prevents initiation of DNA replication. Inhibits cell proliferation in a variety of human cell lines and induces apoptosis in a p53-independent manner in vivo.
Specifications

Chemical Identifiers

942425-68-5
249.698
pha 767491 hydrochloride, pha-767491 hydrochloride, 1,5,6,7-tetrahydro-2-4-pyridinyl-4h-pyrrolo 3,2-c pyridin-4-one hydrochloride, 2-pyridin-4-yl-1h,5h,6h,7h-pyrrolo 3,2-c pyridin-4-one hydrochloride, cay-10572 hydrochloride, pha-00767491 hydrochloride, pha 767491 hydrochloride hplc, 1,5,6,7-tetrahydro-2-4-pyridinyl-??4h-pyrrolo 3,2-c pyridin-4-one hydrochloride, 2-pyridin-4-yl-1,5,6,7-tetrahydro-4h-pyrrolo 3,2-c pyridin-4-one hydrochloride
2-pyridin-4-yl-1,5,6,7-tetrahydropyrrolo[3,2-c]pyridin-4-one;hydrochloride
C12H12ClN3O
IMVNFURYBZMFDZ-UHFFFAOYSA-N
11715766
C1CNC(=O)C2=C1NC(=C2)C3=CC=NC=C3.Cl

Specifications

50mg
942425-68-5
pha 767491 hydrochloride, pha-767491 hydrochloride, 1,5,6,7-tetrahydro-2-4-pyridinyl-4h-pyrrolo 3,2-c pyridin-4-one hydrochloride, 2-pyridin-4-yl-1h,5h,6h,7h-pyrrolo 3,2-c pyridin-4-one hydrochloride, cay-10572 hydrochloride, pha-00767491 hydrochloride, pha 767491 hydrochloride hplc, 1,5,6,7-tetrahydro-2-4-pyridinyl-??4h-pyrrolo 3,2-c pyridin-4-one hydrochloride, 2-pyridin-4-yl-1,5,6,7-tetrahydro-4h-pyrrolo 3,2-c pyridin-4-one hydrochloride
C1CNC(=O)C2=C1NC(=C2)C3=CC=NC=C3.Cl
249.698
249.7
PHA 767491 hydrochloride
C12H12ClN3O
IMVNFURYBZMFDZ-UHFFFAOYSA-N
2-pyridin-4-yl-1,5,6,7-tetrahydropyrrolo[3,2-c]pyridin-4-one;hydrochloride
11715766
>99%
Videos
Documents