missing translation for 'onlineSavingsMsg'
Learn More

PHA 665752, Tocris Bioscience™

Potent and selective MET inhibitor

Brand:  Tocris (Bio-Techne) 269350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13441744

  • £1053.39 / 50mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, selective and ATP-competitive inhibitor of MET kinase (IC50 values are 9, 68, 200, 1400, 3000, 3800 and 6000 nM for MET, Ron, Flk-1, c-abl, FGFR1, EGFR and c-src respectively and > 10000 nM for IGF-IR, PDGFR, AURORA2, PKA, PKBα, p38α, MK2 and MK3). Antitumor agent; inhibits tumorigenicity and angiogenesis in mouse lung cancer xenografts.
Specifications

Chemical Identifiers

477575-56-7
641.608
unii-0vxu5t5r3j, 2r-1-5-z-5-2,6-dichlorophenyl methyl sulfonyl-1,2-dihydro-2-oxo-3h-indol-3-ylidene methyl-2,4-dimethyl-1h-pyrrol-3-yl carbonyl-2-1-pyrrolidinylmethyl pyrrolidine, 0vxu5t5r3j, r,z-5-2,6-dichlorobenzylsulfonyl-3-3,5-dimethyl-4-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylene indolin-2-one, pha-665752 hydrate, 3z-5-2,6-dichlorophenyl methanesulfonyl-3-3,5-dimethyl-4-2r-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylidene-1h-indol-2-one, 3z-5-2,6-dichlorophenyl methylsulfonyl-3-3,5-dimethyl-4-2r-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylidene-1h-indol-2-one, r,z-5-2,6-dichlorobenzyl sulfonyl-3-3,5-dimethyl-4-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylene indolin-2-one, 3z-5-2,6-dichlorophenyl methyl sulfonyl-3-3,5-dimethyl-4-2r-2-1-pyrrolidinylmethyl-1-pyrrolidinyl carbonyl-1h-pyrrol-2-yl methylene-1,3-dihydro-2h-indol-2-one hydrate, d00tbd
(3Z)-5-[(2,6-dichlorophenyl)methylsulfonyl]-3-[[3,5-dimethyl-4-[(2R)-2-(pyrrolidin-1-ylmethyl)pyrrolidine-1-carbonyl]-1H-pyrrol-2-yl]methylidene]-1H-indol-2-one
C32H34Cl2N4O4S
OYONTEXKYJZFHA-SSHUPFPWSA-N
10461815
CC1=C(NC(=C1C(=O)N2CCCC2CN3CCCC3)C)C=C4C5=C(C=CC(=C5)S(=O)(=O)CC6=C(C=CC=C6Cl)Cl)NC4=O

Specifications

PHA 665752
477575-56-7
unii-0vxu5t5r3j, 2r-1-5-z-5-2,6-dichlorophenyl methyl sulfonyl-1,2-dihydro-2-oxo-3h-indol-3-ylidene methyl-2,4-dimethyl-1h-pyrrol-3-yl carbonyl-2-1-pyrrolidinylmethyl pyrrolidine, 0vxu5t5r3j, r,z-5-2,6-dichlorobenzylsulfonyl-3-3,5-dimethyl-4-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylene indolin-2-one, pha-665752 hydrate, 3z-5-2,6-dichlorophenyl methanesulfonyl-3-3,5-dimethyl-4-2r-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylidene-1h-indol-2-one, 3z-5-2,6-dichlorophenyl methylsulfonyl-3-3,5-dimethyl-4-2r-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylidene-1h-indol-2-one, r,z-5-2,6-dichlorobenzyl sulfonyl-3-3,5-dimethyl-4-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylene indolin-2-one, 3z-5-2,6-dichlorophenyl methyl sulfonyl-3-3,5-dimethyl-4-2r-2-1-pyrrolidinylmethyl-1-pyrrolidinyl carbonyl-1h-pyrrol-2-yl methylene-1,3-dihydro-2h-indol-2-one hydrate, d00tbd
CC1=C(NC(=C1C(=O)N2CCCC2CN3CCCC3)C)C=C4C5=C(C=CC(=C5)S(=O)(=O)CC6=C(C=CC=C6Cl)Cl)NC4=O
641.608
641.61
50mg
C32H34Cl2N4O4S
OYONTEXKYJZFHA-SSHUPFPWSA-N
(3Z)-5-[(2,6-dichlorophenyl)methylsulfonyl]-3-[[3,5-dimethyl-4-[(2R)-2-(pyrrolidin-1-ylmethyl)pyrrolidine-1-carbonyl]-1H-pyrrol-2-yl]methylidene]-1H-indol-2-one
10461815
>98%
Videos