missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 7.00 Color-Coded Buffer Solution at 20°C, Reagecon
Easy-to-use, tested, certified, NIST traceable, and shelf-stable, color-coded buffer solution.
Brand: Reagecon 5070C
Code : NEW
Additional Details : Weight : 4.63500kg
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
- NIST traceable
Specifications
Yellow | |
7.00 ±0.01 | |
5 L |
Accuracy ± 0.01 pH units | |
Liquid | |
HDPE Bottle |