missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 6.80 Buffer Solution at 25°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
Brand: Reagecon 1068525
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Accuracy ± 0.01 pH units | |
Liquid | |
HDPE Bottle GL45 |
6.80 ±0.01 | |
500 mL |