Learn More
Invitrogen™ PGF Human ELISA Kit
Sandwich ELISA Kit
£544.00 - £2320.00
Specifications
Accession Number | P49763 |
---|---|
Assay Sensitivity | 2 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465103
|
Invitrogen™
EHPGF |
96 Tests |
£544.00
1 set |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17165539
|
Invitrogen™
EHPGFX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
Description
The Human Placental Growth Factor (PLGF) (Hu PGF) ELISA quantitates Hu PGF in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu PGF. Principle of the method The Human PGF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Placental growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family. PlGF is a pleiotropic factor affects different cell types and regulates various biological responses. The activity of vascular endothelial growth factor (VEGF), which is the most potent of all angiogenic mediators, is partly modulated by PlGF. It plays a major role in vasculogenesis and angiogenesis in human placenta.Specifications
P49763 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
D12S1900L,PIGF,PLGF,PlGF-2,SHGC-10760,PGF | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
2 pg/mL | |
Biotin | |
Growth Factors & Receptors | |
This ELISA kit shows no cross-reactivity with any of the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, MMP-1, - 2, -3, -10, PARC, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
5228 | |
<12% | |
HRP | |
Angiogenesis | |
D12S1900L, PLGF, PlGF-2, SHGC-10760, PGF | |
1 hr. 20 min. |