missing translation for 'onlineSavingsMsg'
Learn More

PG-9 maleate, Tocris Bioscience™

Presynaptic cholinergic modulator

Brand:  Tocris (Bio-Techne) 075050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13427913

  • £292.53 / 50mg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A potent analgesic and nootropic agent. Increases release of acetylcholine.
Specifications

Chemical Identifiers

155649-00-6
468.344
Tropanyl 2-(4-bromophenyl)propionate maleate
(Z)-but-2-enedioic acid;[(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 2-(4-bromophenyl)propanoate
C21H26BrNO6
VBUSCULTLLOSLM-SGCJHOTMSA-N
56972155
CC(C1=CC=C(C=C1)Br)C(=O)OC2CC3CCC(C2)N3C.C(=CC(=O)O)C(=O)O

Specifications

50mg
155649-00-6
Tropanyl 2-(4-bromophenyl)propionate maleate
CC(C1=CC=C(C=C1)Br)C(=O)OC2CC3CCC(C2)N3C.C(=CC(=O)O)C(=O)O
468.344
468.34
PG-9 maleate
C21H26BrNO6
VBUSCULTLLOSLM-SGCJHOTMSA-N
(Z)-but-2-enedioic acid;[(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 2-(4-bromophenyl)propanoate
56972155
>98%
Videos
Documents