missing translation for 'onlineSavingsMsg'
Learn More

Pfu DNA Polymerase

Optimize PCR applications requiring high fidelity with this highly thermostable DNA polymerase.

Brand:  Thermo Scientific EP0502

Code : 88

Additional Details : Weight : 0.03000kg

 View more versions of this product

Product Code. 10202830

  • £223.00 / 500 units


Estimated Shipment
09-02-2018
Add to basket

Description

Description

Highlights

  • Eight times more accurate than Taq DNA polymerase
  • Highly thermostable – remains 95% active after 2 hours incubation at 95°C
  • Generates blunt-end PCR products
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescently-labeled nucleotides)
  • Increased PCR yields with native Pfu DNA Polymerase
Specifications

Specifications

2 x 1.25mL 10X Pfu Buffer with (NH4)2SO2, 2 x 1.25mL 10X Pfu Buffer with MgSO4 and 2 x 1.25mL 25mM MgSO4
Pfu DNA Polymerase (recombinant)
500U
remains 95% active after 2 hours incubation at 95°C
Videos