missing translation for 'onlineSavingsMsg'
Learn More

PF 750, Tocris Bioscience™

Selective FAAH inhibitor

Brand:  Tocris (Bio-Techne) 330750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13462754

  • £705.24 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Irreversible fatty acid amide hydrolase (FAAH) inhibitor (IC50 = 16.2 nM) that displays no activity at a range of other serine hydrolases. Selectively inhibits FAAH within the central nervous system. Orally active.
Specifications

Chemical Identifiers

959151-50-9
345.446
n-phenyl-4-quinolin-3-ylmethyl piperidine-1-carboxamide, unii-7756cpp14k, n-phenyl-4-3-quinolinylmethyl-1-piperidinecarboxamide, d0w0se, 1-piperidinecarboxamide, n-phenyl-4-3-quinolinylmethyl, pf hplc, n-phenyl-4-quinolin-3-yl methyl piperidine-1-carboxamide
N-phenyl-4-(quinolin-3-ylmethyl)piperidine-1-carboxamide
C22H23N3O
BIODYGOZWZNCAG-UHFFFAOYSA-N
25154868
C1CN(CCC1CC2=CC3=CC=CC=C3N=C2)C(=O)NC4=CC=CC=C4

Specifications

PF 750
959151-50-9
n-phenyl-4-quinolin-3-ylmethyl piperidine-1-carboxamide, unii-7756cpp14k, n-phenyl-4-3-quinolinylmethyl-1-piperidinecarboxamide, d0w0se, 1-piperidinecarboxamide, n-phenyl-4-3-quinolinylmethyl, pf hplc, n-phenyl-4-quinolin-3-yl methyl piperidine-1-carboxamide
C1CN(CCC1CC2=CC3=CC=CC=C3N=C2)C(=O)NC4=CC=CC=C4
345.446
345.44
50mg
C22H23N3O
BIODYGOZWZNCAG-UHFFFAOYSA-N
N-phenyl-4-(quinolin-3-ylmethyl)piperidine-1-carboxamide
25154868
>98%
Videos