missing translation for 'onlineSavingsMsg'
Learn More

PF 4800567 hydrochloride, Tocris Bioscience™

Selective casein kinase 1ε inhibitor

Brand:  RD Systems (Bio-Techne) 4281/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13464484

  • £560.32 / 50mg


Estimated Shipment
03-04-2019
Add to basket

Description

Description

Selective casein kinase 1ε inhibitor; displays 22-fold greater potency towards CK1ε than CK1δ (IC 50 values are 32 and 711 nM for CK1ε and CK1δ respectively). ATP competitive. Displays minimal effect on the circadian clock.
Specifications

Chemical Identifiers

1391052-28-0
396.272
pf 4800567 hydrochloride, pf-4800567 hydrochloride, 3-3-chlorophenoxy methyl-1-tetrahydro-2h-pyran-4-yl-1h-pyrazolo 3,4-d pyrimidin-4-amine hydrochloride, 3-3-chlorophenoxymethyl-1-oxan-4-yl pyrazolo 3,4-d pyrimidin-4-amine hydrochloride
3-[(3-chlorophenoxy)methyl]-1-(oxan-4-yl)pyrazolo[3,4-d]pyrimidin-4-amine;hydrochloride
C17H19Cl2N5O2
QZXZQMUZEHTFHD-UHFFFAOYSA-N
71751553
C1COCCC1N2C3=C(C(=N2)COC4=CC(=CC=C4)Cl)C(=NC=N3)N.Cl

Specifications

PF 4800567 hydrochloride
1391052-28-0
pf 4800567 hydrochloride, pf-4800567 hydrochloride, 3-3-chlorophenoxy methyl-1-tetrahydro-2h-pyran-4-yl-1h-pyrazolo 3,4-d pyrimidin-4-amine hydrochloride, 3-3-chlorophenoxymethyl-1-oxan-4-yl pyrazolo 3,4-d pyrimidin-4-amine hydrochloride
C1COCCC1N2C3=C(C(=N2)COC4=CC(=CC=C4)Cl)C(=NC=N3)N.Cl
396.272
396.27
50mg
C17H19Cl2N5O2
QZXZQMUZEHTFHD-UHFFFAOYSA-N
3-[(3-chlorophenoxy)methyl]-1-(oxan-4-yl)pyrazolo[3,4-d]pyrimidin-4-amine;hydrochloride
71751553
>99%
Videos