missing translation for 'onlineSavingsMsg'
Learn More

PF 3845, Tocris Bioscience™

Selective FAAH inhibitor

Brand:  Tocris (Bio-Techne) 417510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13464284

  • £178.20 / 10mg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

Selective fatty acid amide hydrolase (FAAH) inhibitor (K i = 0.23 μM). Reduces inflammatory pain via a cannabinoid receptor-dependent mechanism. Highly efficacious and selective in vivo. Displays no activity at FAAH-2 (IC 50 >10 μM).
Specifications

Chemical Identifiers

1196109-52-0
456.469
unii-q3pw846tyn, n-pyridin-3-yl-4-3-5-trifluoromethyl pyridin-2-yl oxy benzyl piperidine-1-carboxamide, n-3-pyridinyl-4-3-5-trifluoromethyl-2-pyridinyl oxy phenyl methyl-1-piperidinecarboxamide, q3pw846tyn, n-pyridin-3-yl-4-3-5-trifluoromethyl pyridin-2-yl oxy phenyl methyl piperidine-1-carboxamide, pf 3845 hydrate, n-3-pyridinyl-4-3-5-trifluoromethyl-2-pyridinyl oxy phenyl methyl-1-piperidinecarboxamide hydrate, d0e9ux, 1-piperidinecarboxamide, n-3-pyridinyl-4-3-5-trifluoromethyl-2-pyridinyl oxy phenyl methyl, n-3-pyrdnyl-4-3-5-trfluoromethyl-2-pyrdnyl oxy phenyl methyl-1-pperdnecarboxamde
N-pyridin-3-yl-4-[[3-[5-(trifluoromethyl)pyridin-2-yl]oxyphenyl]methyl]piperidine-1-carboxamide
C24H23F3N4O2
NBOJHRYUGLRASX-UHFFFAOYSA-N
25154867
C1CN(CCC1CC2=CC(=CC=C2)OC3=NC=C(C=C3)C(F)(F)F)C(=O)NC4=CN=CC=C4

Specifications

PF 3845
1196109-52-0
unii-q3pw846tyn, n-pyridin-3-yl-4-3-5-trifluoromethyl pyridin-2-yl oxy benzyl piperidine-1-carboxamide, n-3-pyridinyl-4-3-5-trifluoromethyl-2-pyridinyl oxy phenyl methyl-1-piperidinecarboxamide, q3pw846tyn, n-pyridin-3-yl-4-3-5-trifluoromethyl pyridin-2-yl oxy phenyl methyl piperidine-1-carboxamide, pf 3845 hydrate, n-3-pyridinyl-4-3-5-trifluoromethyl-2-pyridinyl oxy phenyl methyl-1-piperidinecarboxamide hydrate, d0e9ux, 1-piperidinecarboxamide, n-3-pyridinyl-4-3-5-trifluoromethyl-2-pyridinyl oxy phenyl methyl, n-3-pyrdnyl-4-3-5-trfluoromethyl-2-pyrdnyl oxy phenyl methyl-1-pperdnecarboxamde
C1CN(CCC1CC2=CC(=CC=C2)OC3=NC=C(C=C3)C(F)(F)F)C(=O)NC4=CN=CC=C4
456.469
456.46
10mg
C24H23F3N4O2
NBOJHRYUGLRASX-UHFFFAOYSA-N
N-pyridin-3-yl-4-[[3-[5-(trifluoromethyl)pyridin-2-yl]oxyphenyl]methyl]piperidine-1-carboxamide
25154867
>99%
Videos