missing translation for 'onlineSavingsMsg'
Learn More

PF 04217903 mesylate, Tocris Bioscience™

Highly selective MET inhibitor

Brand:  Tocris (Bio-Techne) 423910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13444414

  • £255.78 / 10mg

Estimated Shipment date
from Supplier 28-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly selective, high affinity MET inhibitor (Ki = 6-7 nM against wild type c-Met). Displays >1000-fold selectivity for c-Met over a panel of 208 kinases.
Specifications

Chemical Identifiers

956906-93-7
468.492
pf 04217903 mesylate, pf-04217903 methanesulfonate, pf-04217903 mesylate, 2-4-3-quinolin-6-ylmethyl-1,2,3 triazolo 4,5-b pyrazin-5-yl pyrazol-1-yl ethanol; methanesulfonic acid, c19h16n8o.ch4o3s, 1h-pyrazole-1-ethanol, 4-1-6-quinolinylmethyl-1h-1,2,3-triazolo 4,5-b pyrazin-6-yl-, methanesulfonate 1:1, 2-4-1-quinolin-6-ylmethyl-1h-1,2,3 triazolo 4,5-b pyrazin-6-yl-1h-pyrazol-1-yl ethanol methanesulfonate 1:1, pf-04217903 mesylate/pf04217903 mesylate, pf hplc
methanesulfonic acid;2-[4-[3-(quinolin-6-ylmethyl)triazolo[4,5-b]pyrazin-5-yl]pyrazol-1-yl]ethanol
C20H20N8O4S
HBEMHKVWZJTVOC-UHFFFAOYSA-N
24852079
CS(=O)(=O)O.C1=CC2=C(C=CC(=C2)CN3C4=NC(=CN=C4N=N3)C5=CN(N=C5)CCO)N=C1

Specifications

10mg
956906-93-7
pf 04217903 mesylate, pf-04217903 methanesulfonate, pf-04217903 mesylate, 2-4-3-quinolin-6-ylmethyl-1,2,3 triazolo 4,5-b pyrazin-5-yl pyrazol-1-yl ethanol; methanesulfonic acid, c19h16n8o.ch4o3s, 1h-pyrazole-1-ethanol, 4-1-6-quinolinylmethyl-1h-1,2,3-triazolo 4,5-b pyrazin-6-yl-, methanesulfonate 1:1, 2-4-1-quinolin-6-ylmethyl-1h-1,2,3 triazolo 4,5-b pyrazin-6-yl-1h-pyrazol-1-yl ethanol methanesulfonate 1:1, pf-04217903 mesylate/pf04217903 mesylate, pf hplc
CS(=O)(=O)O.C1=CC2=C(C=CC(=C2)CN3C4=NC(=CN=C4N=N3)C5=CN(N=C5)CCO)N=C1
468.492
468.49
PF 04217903 mesylate
C20H20N8O4S
HBEMHKVWZJTVOC-UHFFFAOYSA-N
methanesulfonic acid;2-[4-[3-(quinolin-6-ylmethyl)triazolo[4,5-b]pyrazin-5-yl]pyrazol-1-yl]ethanol
24852079
>98%
Videos
Documents