missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, Reagent ACS, Thermo Scientific Chemicals

Catalog Number p-216706
20.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 mL
5 L
Packaging:
EcoSafPak™ Glass Bottle
Plastic Jerrycan
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
64742-49-0
C6H14
86.178
MFCD00081849
AFABGHUZZDYHJO-UHFFFAOYSA-N
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
7892
Petroleum ether
CCCC(C)C
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
64742-49-0
C6H14
86.178
MFCD00081849
AFABGHUZZDYHJO-UHFFFAOYSA-N
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
7892
Petroleum ether
CCCC(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity Passes Test
Density 0.6480g/mL
Boiling Point 35°C to 60°C
Quantity 500 mL
Flash Point −30°C (-86°F)
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max.
UN Number 1268
Merck Index 14, 5489
Grade ACS Reagent
Chemical Name or Material Petroleum Ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'