missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether 35/60, Environmental Grade, Alfa Aesar™

Suitable For Environmental and Pesticide Analysis

Brand:  Alfa Aesar 040982.M1

Code : NEW

Additional Details : CAS Number : 8032-32-4 Weight : 0.01000kg Transport : UN number : 1268 Chem class : 1 Pack group : I

Quantity 4 x 1L

 View more versions of this product

Product Code. 11395317

  • £73.40 / 4 L


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Specifications

Chemical Identifiers

Petroleum ether 35/60
8032-32-4
Liquid
MFCD00081849
Environmental Grade
Environmental Grade
>98%

Specifications

Petroleum ether 35/60
8032-32-4
Colorless liquid
>98%
250°C (482°F)
350 hPa (263mm Hg) at 20°C
4 x 1L
Environmental Grade
Environmental Grade
Liquid
MFCD00081849
Petroleum-like
Clear colorless
Videos
SDS