missing translation for 'onlineSavingsMsg'
Learn More

Westfield Medical Sterilisation pouch gussetted, chevron shaped end with thumb holes 300mm x 65mm x 500mm

Brand:  Westfield Medical WPG300X500

Sterilisation pouch gussetted, chevron shaped end with thumb holes 300mm x 65mm x 500mm

Code : 03

Additional Details : Weight : 0.20000kg

Product Code. 12769035

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.