0 PET/PP Film Pouch
Overview
Brand: 0
Manufacturer Part Number: WP100X300
UNSPSC: 42281904
Code: 03
Additional Details:
Additional Details: Weight: 3.70000kg
Product Code. 12769015
Quantity Price
1 £ 112.0 / Pack of 1000
Estimated Shipment
28-04-2017
Add to basket
Description and Specification
Specification
Material Polyethylene terephthalate/Polypropylene Film
Certifications/Compliance CE Marked
Quantity 1000Pack
For Use With (Application) Suitable For Steam and ETO Sterilisation, amd formaldehyde