0 PET/PP Film Pouch
Overview
Brand: 0
Manufacturer Part Number: WP150X200
X1000 Sterilisation pouch flat, chevron shaped
UNSPSC: 42281904
Code: 03
Additional Details:
Additional Details: Weight: 0.20000kg
Product Code. 12709025
Quantity Price
1 £ 143.0 / Pack of 1000
Estimated Shipment
08-02-2017
Add to basket
Description and Specification
Specification
Packaging 1000Pack
Material Polyethylene terephthalate/Polypropylene Film
For Use With (Application) Suitable For Steam and ETO Sterilisation, amd formaldehyde
Certifications/Compliance CE Marked