missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Pestle

Brand:  Haldenwanger™ 56/00/rauh

Code : 02

Additional Details : Weight : 0.06500kg

 View more versions of this product

Product Code. 12091802

  • £9.89 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

24mm
Pestle
115mm
Pestle
Videos