missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Pestle

Brand:  Haldenwanger™ 56/0a/glasiert

UNSPSC : 41120000

Code : 02

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12001812

  • £26.15 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

Pestle
Pestle
28mm
125mm