missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Pestle

Brand:  Haldenwanger™ 56/3/glasiert

UNSPSC : 41120000

Code : 02

Additional Details : Weight : 0.21000kg

 View more versions of this product

Product Code. 12061812

  • £43.40 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

42mm
Pestle
175mm
Pestle