missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Pestle

Brand:  Haldenwanger™ 56/4/rauh

Code : 02

Additional Details : Weight : 0.23000kg

 View more versions of this product

Product Code. 12091812

  • £24.81 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

45mm
Pestle
180mm
Pestle
Videos