missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Pestle

Brand:  Haldenwanger™ 56-6A

Code : 02

Additional Details : Weight : 0.29516kg

 View more versions of this product

Product Code. 11347454

  • £39.82 / Each


Estimated Shipment
28-06-2018
Add to basket

Description

Description

Grinding surfaces are unglazed.
Specifications

Specifications

55mm
Pestle
210mm
Pestle
Videos
Documentation View accessory products