missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Pestle

Brand:  Haldenwanger™ 56/8/glasiert

Code : 02

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10507654

  • £49.86 / Each


Estimated Shipment
19-02-2018
Add to basket

Specifications

Specifications

60mm
Pestle
215mm
Pestle
Videos