Learn More
Perforin, PE-Cyanine7, clone: dG9 (delta G9), eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 25-9994-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Perforin is one of the major cytolytic proteins of cytolytic granules. One of the main pathways of lymphocyte mediated cytolysis entails the secretion onto target membranes of cytolytic granules contained in cytolytic effector lymphocytes of T-cell or NK-cell type. Perforin is a cytolytic mediator and is stored in and released by cytoplasmic granules. Perforin (PRF) is involved in the killing mediated by cytotoxic lymphocytes which represents an important mechanism in the immune defense against tumors and virus infections. Human PRF is a 555 amino acid protein with a 21 amino acid signal peptide. It has a molecular weight of 70 to 75 kD. PRF is a pore forming protein with a mechanism of transmembrane channel formation similar to C9 and homology between perforin and C9 has been demonstrated at their respective functionally conserved regions. Studies show that perforin is expressed only in killer cell lines and not in helper T lymphocytes or other tumor cells tested.
Description: The dG9 antibody clone reacts with human perforin (pore-forming protein, pfp). Perforin is one of the cytolytic mediators present in the cytoplasmic granules of cytotoxic T lymphocytes (CTL) and natural killer cells (NK). Perforin is involved in the killing function by CTLs and NKs and has an important role in the immune response against tumors and virus infections.Applications Reported: This dG9 (delta G9) antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This dG9 (delta G9) antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
Perforin | |
Monoclonal | |
5μl/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P14222 | |
PRF1 | |
Affinity chromatography | |
RUO | |
396595, 5551 | |
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures. | |
Liquid |
Flow Cytometry | |
dG9 (delta G9) | |
PE-Cyanine7 | |
PRF1 | |
cytolysin, FLH2, HPLH2, lymphocyte pore forming protein, lymphocyte pore-forming protein, perforin 1 (pore forming protein), perforin-1, PFN1, PFP, RP11-710A11.3 | |
Mouse | |
25 Tests | |
Primary | |
Human, Porcine | |
Antibody | |
IgG2b κ |
For Research Use Only.