missing translation for 'onlineSavingsMsg'
Learn More

Perforin Mouse anti-Human, Porcine, eFluor® 450, Clone: dG9 (delta G9), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-9994-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15599856

  • £200.00 / 100 tests

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered. Perforin is one of the major cytolytic proteins of cytolytic granules. Perforin is a cytolytic mediator and is stored in and released by cytoplasmic granules. Moreover, perforin is involved in immune defense against tumors and virus infections as mediated by cytotoxic lymphocytes. Perforin is a 555 amino acid protein with a 21 amino acid signal peptide, and has a molecular weight of 70 to 75 kD. Perforin is a pore forming protein with a mechanism of transmembrane channel formation similar to C9, and homology between perforin and C9 have been demonstrated. Studies show that perforin is expressed only in killer cell lines and not in helper T lymphocytes or other tumor cells tested. Perforin is known to be a key effector molecule for T-cell- and natural killer-cell-mediated cytolysis. Defects in the perforin gene cause familial hemophagocytic lymphohistiocytosis type 2 (HPLH2), a rare and lethal autosomal recessive disorder of early childhood. Alternative splicing results in multiple transcript variants of perforin.
Specifications

Specifications

Perforin
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P14222
cytolysin; FLH2; HPLH2; lymphocyte pore forming protein; lymphocyte pore-forming protein; perforin 1 (pore forming protein); perforin-1; PFN1; PFP; RP11-710A11.3
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Porcine
Flow Cytometry
dG9 (delta G9)
eFluor™ 450
PRF1
Liquid
PRF1
IgG2b, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
396595, 5551
Videos
SDS
Documents

For Research Use Only.