missing translation for 'onlineSavingsMsg'
Learn More

Perfluorotripentylamine, tech. 85%, Thermo Scientific Chemicals

Catalog Number 11313796
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
338-84-1
C15F33N
821.119
MFCD00042367
AQZYBQIAUSKCCS-UHFFFAOYSA-N
perfluorotripentylamine, tris undecafluoropentyl amine, perfluorotri-n-pentylamine, perfluorotriamylamine, fluorinert fc-70, perfluoro-compound fc-70, 1-pentanamine, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis undecafluoropentyl, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis 1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl pentan-1-amine, 1-pentanamine, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis 1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl, fluorinert fc 70
67646
1,1,2,2,3,3,4,4,5,5,5-undecafluoro-N,N-bis(1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl)pentan-1-amine
C(C(C(N(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
338-84-1
C15F33N
821.119
MFCD00042367
AQZYBQIAUSKCCS-UHFFFAOYSA-N
perfluorotripentylamine, tris undecafluoropentyl amine, perfluorotri-n-pentylamine, perfluorotriamylamine, fluorinert fc-70, perfluoro-compound fc-70, 1-pentanamine, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis undecafluoropentyl, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis 1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl pentan-1-amine, 1-pentanamine, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis 1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl, fluorinert fc 70
67646
1,1,2,2,3,3,4,4,5,5,5-undecafluoro-N,N-bis(1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl)pentan-1-amine
C(C(C(N(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.94
Boiling Point 210°C to 220°C
Quantity 5 g
Formula Weight 821.12
Percent Purity 85%
Grade Technical
Chemical Name or Material Perfluorotripentylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'