missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluoroheptanoic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Perfluoroheptanoic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Perfluoroheptanoic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Perfluoroheptanoic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Melting Point | 31°C to 36°C (literature) |
Boiling Point | 89°C (30 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 100 g |
UN Number | UN3261 |
Beilstein | 1808210 |
Formula Weight | 364.06 |
Percent Purity | ≥98% |
Chemical Name or Material | Perfluoroheptanoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'