missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1-iodohexane, 97%, ACROS Organics™

Brand:  Acros Organics 269941000

Code : NEW

Additional Details : CAS Number : 355-43-1 Weight : 0.10000kg


Quantity 100g

 View more versions of this product

Product Code. 15402142

  • £257.76 / 100g

Estimated Shipment: 09-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

355-43-1
445.95
perfluorohexyl iodide, perfluoro-1-iodohexane, 1-iodoperfluorohexane, perfluoro-n-hexyl iodide, tridecafluorohexyl iodide, unii-a2a17jug4r, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodo, tridecafluoro-1-iodohexane, a2a17jug4r, hexane, tridecafluoro-6-iodo
1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
C6F13I
BULLJMKUVKYZDJ-UHFFFAOYSA-N
67733
C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Perfluoro-1-iodohexane
>115°C
1.3270 to 1.3300
116.0°C to 119.0°C
100g
96% min. (GC)
CF3(CF2)5I
Solubility in water: insoluble.
C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
445.95
445.95
2.0630g/mL
Authentic
2.063
-45.0°C
355-43-1
C6F13I
perfluorohexyl iodide, perfluoro-1-iodohexane, 1-iodoperfluorohexane, perfluoro-n-hexyl iodide, tridecafluorohexyl iodide, unii-a2a17jug4r, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodo, tridecafluoro-1-iodohexane, a2a17jug4r, hexane, tridecafluoro-6-iodo
BULLJMKUVKYZDJ-UHFFFAOYSA-N
1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
67733
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Perfluoro-1-iodohexane

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents
Certificates