missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloromethyl mercaptan, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Density | 1.7000g/mL |
Boiling Point | 146.0°C to 148.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Linear Formula | CCl3SCl |
Refractive Index | 1.5370 to 1.5410 |
Quantity | 25 g |
Beilstein | 03,135 |
Show More |
Safety and Handling
Product Identifier
- Perchloromethyl mercaptan
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 1
- H301-Toxic if swallowed.
- H330-Fatal if inhaled.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'