missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 70.0-72.0%, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Concentration | 70 to 72% |
Residue after Ignition | ≤0.003% (as SO4) |
Flash Point | 113°C (235.4°F) |
Quantity | 500 mL |
Linear Formula | HClO4 |
UN Number | 1873 |
Formula Weight | 100.46g/mol |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Perchloric Acid, Reag. Ph. Eur. reag. ISO |
Safety and Handling
Product Identifier
- Perchloric acid
- Danger
- Oxidising solid Category 1
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- H271-May cause fire or explosion; strong oxidiser.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'